Skip to Content
Merck
1-Ethyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide ≥99%, H2O <500 ppm

1-Ethyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide

1-Ethyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide ≥99%, H2O <500 ppm
Empirical Formula (Hill Notation):
C9H16F6N2O4S2
CAS Number:
Molecular Weight:
394.35

Products

Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group