![1-Cyclopropyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/200/847/9d4d6cb5-9b73-4ed1-9d43-f0e69d126252/640/9d4d6cb5-9b73-4ed1-9d43-f0e69d126252.png)
1-Cyclopropyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one
![1-Cyclopropyl-6-fluoro-7-(piperazin-1-yl)quinolin-4(1H)-one certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/200/847/9d4d6cb5-9b73-4ed1-9d43-f0e69d126252/640/9d4d6cb5-9b73-4ed1-9d43-f0e69d126252.png)
Synonym(s):
Decarboxy Ciprofloxacin, MBX 2192
Empirical Formula (Hill Notation):
C16H18FN3O
CAS Number:
Molecular Weight:
287.33
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1