![1-Bromo-2,4-dimethoxybenzene 97%](/deepweb/assets/sigmaaldrich/product/structures/154/911/5537bb5d-67ca-4e99-bbc7-842572528bb7/640/5537bb5d-67ca-4e99-bbc7-842572528bb7.png)
1-Bromo-2,4-dimethoxybenzene
![1-Bromo-2,4-dimethoxybenzene 97%](/deepweb/assets/sigmaaldrich/product/structures/154/911/5537bb5d-67ca-4e99-bbc7-842572528bb7/640/5537bb5d-67ca-4e99-bbc7-842572528bb7.png)
Linear Formula:
BrC6H3(OCH3)2
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing