![1,3-Diaminopropane dihydrochloride 98%](/deepweb/assets/sigmaaldrich/product/structures/422/487/84a918de-3d50-4adb-9bee-288773e5b6e4/640/84a918de-3d50-4adb-9bee-288773e5b6e4.png)
1,3-Diaminopropane dihydrochloride
![1,3-Diaminopropane dihydrochloride 98%](/deepweb/assets/sigmaaldrich/product/structures/422/487/84a918de-3d50-4adb-9bee-288773e5b6e4/640/84a918de-3d50-4adb-9bee-288773e5b6e4.png)
Synonym(s):
1,3-Propanediamine, 1,3-Propanediamine dihydrochloride
Linear Formula:
H2N(CH2)3NH2·2HCl
CAS Number:
Molecular Weight:
147.05
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing