Skip to Content
Merck
All Photos(1)

Key Documents

F19353

Sigma-Aldrich

Fumaric acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH=CHCOOH
CAS Number:
Molecular Weight:
116.07
Beilstein:
605763
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021709
PubChem Substance ID:
Pricing and availability is not currently available.

vapor pressure

1.7 mmHg ( 165 °C)

Assay

99%

form

powder

autoignition temp.

1364 °F

expl. lim.

40 %

mp

298-300 °C (subl.) (lit.)

SMILES string

OC(=O)\C=C\C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+

InChI key

VZCYOOQTPOCHFL-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

523.4 °F

Flash Point(C)

273 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tahar Ghnaya et al.
Chemosphere, 90(4), 1449-1454 (2012-10-03)
The implication of organic acids in Pb translocation was studied in two species varying in shoot lead accumulation, Sesuvium portulacastrum and Brassica juncea. Citric, fumaric, malic and α-cetoglutaric acids were separated and determined by HPLC technique in shoots, roots and
Emine Akar et al.
Carbohydrate polymers, 90(4), 1634-1641 (2012-09-05)
A novel biodegradable sodium carboxymethyl cellulose (NaCMC)-based hydrogel was synthesized by using fumaric acid (FA) as a crosslinking agent at various ratios. Hydrogels (CMCF) were characterized using Fourier transform infrared spectroscopy (FTIR), thermogravimetric analysis (TGA), differential scanning calorimetry (DSC), X-ray
Bas J Meussen et al.
Applied microbiology and biotechnology, 94(4), 875-886 (2012-04-25)
Rhizopus oryzae is a filamentous fungus belonging to the Zygomycetes. It is among others known for its ability to produce the sustainable platform chemicals L: -(+)-lactic acid, fumaric acid, and ethanol. During glycolysis, all fermentable carbon sources are metabolized to
Timothy E L Douglas et al.
Macromolecular bioscience, 12(8), 1077-1089 (2012-06-01)
Alkaline phosphatase (ALP), an enzyme involved in mineralization of bone, is incorporated into three hydrogel biomaterials to induce their mineralization with calcium phosphate (CaP). These are collagen type I, a mussel-protein-inspired adhesive consisting of PEG substituted with catechol groups, cPEG
Rohan A Davis et al.
Bioorganic & medicinal chemistry, 18(1), 14-18 (2009-12-08)
We have investigated the enzyme inhibition characteristics of a natural product (NP)-based phenolic library against a panel of human carbonic anhydrases (hCAs, EC 4.2.1.1) which included hCAs I and II (cytosolic) and hCA VA/VB (mitochondrial isoforms). Most of these compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service