Skip to Content
Merck
All Photos(1)

Key Documents

HG000C24

Millipore

Nutrient Broth

pH 7.1±0.2 (25 °C, after autoclaving)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

composition

Meat Extract, 1.0 g/L
Peptone, 5.0 g/L
Sodium Chloride, 8.0 g/L
Yeast Extract, 2.0 g/L

packaging

poly bottle of 12 × 500 g (HG000C24.500)

pH

7.1±0.2 (25 °C, after autoclaving)

storage temp.

15-25°C

Application

A general purpose medium suitable for the cultivation of a wide range of non-fastidious micro-organisms.

Preparation Note

Suspend 16g in 1 litre demineralised water. Mix well and dispense into final containers. Autoclave at 121°C for 15 minutes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service