Skip to Content
Merck
All Photos(1)

Documents

556114

Sigma-Aldrich

N-(4-Carboxyphenyl)succinamic acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO5
CAS Number:
Molecular Weight:
237.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

mp

235-236 °C (lit.)

SMILES string

OC(=O)CCC(=O)Nc1ccc(cc1)C(O)=O

InChI

1S/C11H11NO5/c13-9(5-6-10(14)15)12-8-3-1-7(2-4-8)11(16)17/h1-4H,5-6H2,(H,12,13)(H,14,15)(H,16,17)

InChI key

IOKXKKSBNJCKOY-UHFFFAOYSA-N

Gene Information

human ... BCHE(590)

Application

N-(4-Carboxyphenyl)succinamic acid may be used to synthesize N-(4-carboxyphenyl)succinimide.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antimicrobial activity of new succinimides bearing different heterocycles succinimides bearing different heterocycles.
Al-Azzawi AM and Hassan AS.
International Journal of Research in Pharmacy and Chemistry, 4(4), 755-762 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service