Skip to Content
Merck
All Photos(1)

Key Documents

47727

Sigma-Aldrich

1-Formylpiperazine

technical, ≥90% (GC)

Synonym(s):

Piperazine-1-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O
CAS Number:
Molecular Weight:
114.15
Beilstein:
110295
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

refractive index

n20/D 1.512

bp

264-266 °C

density

1.107 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)N1CCNCC1

InChI

1S/C5H10N2O/c8-5-7-3-1-6-2-4-7/h5-6H,1-4H2

InChI key

MSSDTZLYNMFTKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Formylpiperazine (FPA), a water soluble alicyclic secondary amine, can be prepared by reacting piperazine with methyl formate. It reacts with benzoylisothiocyanate to form 1-formyl-4-(N-benzoylthiocarbamoyl)piperazine. The conformational analysis of FPA based on its vibrational spectral data has been reported. The kinetics of reaction of p-nitrophenyl acetate with FPA in different organic solvents and ionic liquids has been investigated.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics and mechanism of the aminolysis of O-ethyl S-aryl dithiocarbonates.
Oh HK, et al.
The Journal of Organic Chemistry, 56(18), 5324-5328 (1991)
Gürkan Keşan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 118, 1113-1120 (2013-10-29)
Infrared and Raman spectra of 1-formylpiperazine (1-fp) have been recorded in the region of 4000-50 cm(-1). The conformational analysis, optimized geometric parameters, normal mode frequencies and corresponding vibrational assignments of 1-fp (C5H10N2O) have been examined by means of Becke-3-Lee-Yang-Parr (B3LYP)
1-Formylpiperazine and related compounds.
Horrom BW, et al.
Journal of the American Chemical Society, 77(3), 753-754 (1955)
Influence of the ionic liquid on the rate and the mechanism of reaction of p-nitrophenyl acetate with secondary alicyclic amines.
Millan D, et al.
New. J. Chem., 37(10), 3281-3288 (2013)
Thiocarbamoyl derivatives of N-acylpiperazines.
Dhawan B and Southwick PL.
Journal of Heterocyclic Chemistry, 20(1), 243-244 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service