Merck
HomeProfessor Product PortalJamison Group – Professor Product Portal
Jamison Group – Professor Product Portal
Professor Timothy Jamison
Professor Timothy Jamison
Research in the Jamison group is centered on the development of new reactions and technologies for organic synthesis. Towards these themes, the group has pioneered a number of air-stable nickel precatalysts supported by phosphines and N-heterocyclic carbenes that are readily converted to the active catalyst in situ. A variety of carbon-carbon bonding forming reactions promoted by nickel catalysts have also been discovered, including the coupling of different functional groups in a highly regio-, stereo- and enantioselective fashion depending on the nature of the supporting ligands on nickel.
The Jamison group also pursues new chemical methods and technologies under continuous flow conditions to enable reactions that would be difficult or impossible to perform under batch conditions, streamline multistep processes, decrease safety risks to researchers, as well as improve reaction efficiency.
Jamison Group Website
Recent papers from the Jamison Group
1.
Standley EA, Jamison TF. 2013. Simplifying Nickel(0) Catalysis: An Air-Stable Nickel Precatalyst for the Internally Selective Benzylation of Terminal Alkenes. J. Am. Chem. Soc.. 135(4):1585-1592. https://doi.org/10.1021/ja3116718
2.
Standley EA, Smith SJ, Müller P, Jamison TF. 2014. A Broadly Applicable Strategy for Entry into Homogeneous Nickel(0) Catalysts from Air-Stable Nickel(II) Complexes. Organometallics. 33(8):2012-2018. https://doi.org/10.1021/om500156q
3.
Tasker SZ, Standley EA, Jamison TF. 2014. Recent advances in homogeneous nickel catalysis. Nature. 509(7500):299-309. https://doi.org/10.1038/nature13274
Sorry, an unexpected error has occurred
Network error: Failed to fetch
DISCIPLINES
  • Organic Synthetic Chemistry
  • Catalysis
Professor Product Portal Index