Merck
  • Ru(II)/diphenylphosphine/pyridine-6-thiolate complexes induce S-180 cell apoptosis through intrinsic mitochondrial pathway involving inhibition of Bcl-2 and p53/Bax activation.
Ru(II)/diphenylphosphine/pyridine-6-thiolate complexes induce S-180 cell apoptosis through intrinsic mitochondrial pathway involving inhibition of Bcl-2 and p53/Bax activation.
Molecular and cellular biochemistry (2017-08-11)
Wanessa Carvalho Pires, Benedicto Augusto Vieira Lima, Flávia de Castro Pereira, Aliny Pereira Lima, Francyelli Mello-Andrade, Hugo Delleon Silva, Monize Martins da Silva, Legna Colina-Vegas, Javier Ellena, Alzir A Batista, Elisângela de Paul Silveira-Lacerda
ABSTRACT
The aim of this work was the synthesis, characterization, and cytotoxicity evaluation of three new Ru(II) complexes with a general formula [Ru(Spy)(bipy)(P-P)]PF
Sigma-Aldrich
1,3-Bis(diphenylphosphino)propane, 97%