Merck
  • LC-MS/MS determination of d-mannose in human serum as a potential cancer biomarker.
LC-MS/MS determination of d-mannose in human serum as a potential cancer biomarker.
Journal of pharmaceutical and biomedical analysis (2017-01-17)
Lyndsey White, Jing Ma, Su Liang, Beatriz Sanchez-Espiridion, Dong Liang
ABSTRACT
Several metabolites in human serum have been identified as potential cancer biomarkers for early detection. This study focuses on the LC-MS/MS method development and validation of d-mannose in human serum. Surrogate blank serum, coupled with stable isotope d-mannose-13C6, as internal standard, was used for generating standard curves ranging from 1 to 50μg/mL. Separation was achieved by an Agilent 1200 series HPLC equipped with a SUPELCOGELTM Pb, 6% Crosslinked column with HPLC water as a mobile phase at flow rate of 0.5mL/min at 80°C. Mass detection was performed under negative ionization electrospray. Inter- and intra-day accuracy and precision were <2%. The extraction recovery and matrix effect were 104.1%-105.5% and 97.0%-100.0%, respectively. This method was successfully applied for the quantification of d-mannose in the serum samples of 320 esophageal cancer patients and 323 healthy volunteers. We report a simple, specific and reproducible LC-MS/MS method for the quantification of d-mannose in human serum as a potential cancer biomarker.
Product Description
Supelco
SUPELCOGEL Pb, 6% Crosslinked HPLC Column, 9 μm particle size, L × I.D. 30 cm × 7.8 mm
Supelco
SUPELCOGEL Pb, 6% Crosslinked HPLC Column, 9 μm particle size, L × I.D. 10 cm × 7.8 mm
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)