Merck
  • Visible light-induced crosslinking and physiological stabilization of diselenide-rich nanoparticles for redox-responsive drug release and combination chemotherapy.
Visible light-induced crosslinking and physiological stabilization of diselenide-rich nanoparticles for redox-responsive drug release and combination chemotherapy.
Biomaterials (2017-01-10)
Shaodong Zhai, Xianglong Hu, Yongjun Hu, Baoyan Wu, Da Xing
ABSTRACT
Undesired physiological instability of nanocarriers and premature drug leakage during blood circulation result in compromised therapeutic efficacy and severe side effects, which have significantly impeded the development of nanomedicine. Facile crosslinking of drug-loaded nanocarriers while keeping the potency of site-specific degradation and drug release has emerged as a viable strategy to overcome these drawbacks. Additionally, combination therapy has already shown advantages in inhibiting advanced tumors and life extension than single drug therapy. Herein, three kinds of diselenide-rich polymers were fabricated with distinct hydrophobic side chains. The component effect was interrogated to screen out PEG-b-PBSe diblock copolymer due to its favorable self-assembly controllability and high drug loading of camptothecin (CPT) and doxorubicin (DOX) that had synergistic antitumor property. Facile visible light-induced diselenide metathesis and regeneration was employed to crosslink nanocarriers for the first time. The dual drug-loaded crosslinked micelles (CPT/DOX-CCM) were stable in physiological conditions with minimal drug leakage, possessing extended blood circulation, whereas hand-in-hand dual drug release was significantly accelerated in tumor's redox microenvironments. In vitro cytotoxicity evaluation and in vivo tumor suppression with low dosage drugs further demonstrated the favorable potency of the redox-responsive nanoplatform in tumor combination chemotherapy.
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
Sigma-Aldrich
4-Bromobenzyl alcohol, 99%