Merck
  • Synthesis and bio-molecular study of (+)-N-Acetyl-α-amino acid dehydroabietylamine derivative for the selective therapy of hepatocellular carcinoma.
Synthesis and bio-molecular study of (+)-N-Acetyl-α-amino acid dehydroabietylamine derivative for the selective therapy of hepatocellular carcinoma.
BMC cancer (2016-11-16)
Muhammad Ayaz Mustufa, Cigdem Ozen, Imran Ali Hashmi, Afshan Aslam, Jameel Ahmed Baig, Gokhan Yildiz, Shoaib Muhammad, Imam Bakhsh Solangi, Naim Ul Hasan Naqvi, Mehmet Ozturk, Firdous Imran Ali
ABSTRACT
The purpose of present work is to synthesize novel (+)-Dehydroabietylamine derivatives (DAAD) using N-acetyl-α-amino acid conjugates and determine its cytotoxic effects on hepatocellular carcinoma cells. An analytical study was conducted to explore cytotoxic activity of DAAD on hepatocellular carcinoma cell lines. The cytotoxicity effect was recorded using sulforhodamine B technique. Cell cycle analysis was performed using Propidium Iodide (PI) staining. Based on cell morphology, anti growth activity and microarray findings of DAAD2 treatment, Comet assay, Annexin V/PI staining, Immunoperoxidase assay and western blots were performed accoringly. Hep3B cells were found to be the most sensitive with IC It is concluded that a novel DAAD2 with IC
Sigma-Aldrich
(+)-Dehydroabietylamine, technical grade, 60%