- B cell conducts the lymphocyte orchestra.
B cell conducts the lymphocyte orchestra.
The interest for B cells has recently been revived. They normally play a role in the development, the regulation, as well as the activation of lymphoid architecture: they regulate dendritic cells and T-cell subsets function through cytokine production. Receptor editing is also essential in B cells and aids in preventing autoimmunity. Both abnormalities in the distribution of B-cell subsets and clinical benefit response to B-cell depletion in autoimmune states illustrate their importance. A new area has thus been reached, whereby B lymphocytes return as a significant contributor to autoimmune disorders.