Merck
  • Synthesis of (+)-polyoxamic acid and D-sorbitol from simple achiral allylic halides employing (S,S)-hydrobenzoin as a chiral source.
Synthesis of (+)-polyoxamic acid and D-sorbitol from simple achiral allylic halides employing (S,S)-hydrobenzoin as a chiral source.
Chemical communications (Cambridge, England) (2002-07-19)
Kwan Soo Kim, Yong Joo Lee, Jin Hwan Kim, Da Kyung Sung
ABSTRACT
Coupling of cis-1-bromo-2-pentene and (S,S)-hydrobenzoin stannylene acetal followed by regio- and stereoselective transformations of the resulting allylic ether gave (+)-polyoxamic acid and a similar procedure was applied to the synthesis of D-sorbitol from trans-1-iodo-2-hexene.
Sigma-Aldrich
1-Bromo-2-pentene, predominantly trans, 95%