Merck
  • Propargylation of arabinogalactan with propargyl halides--a facile route to new functionalized biopolymers.
Propargylation of arabinogalactan with propargyl halides--a facile route to new functionalized biopolymers.
Carbohydrate research (2013-06-04)
Lyudmila A Grischenko, Lidiya N Parshina, Lyudmila V Kanitskaya, Lyudmila I Larina, Lyubov N Novikova, Boris A Trofimov
ABSTRACT
New arabinogalactan propargyl ethers with degree of substitution (DS) up to 2.8 have been obtained by propargylation of arabino-3,6-galactan (AG) with propargyl bromide (PB) in the two-phase system 30-60% КОН aqueous solution/toluene in the presence of triethylbenzylammonium chloride (TEBAС) or without catalyst (ambient temperature, 1-24 h) in 20-87% yields. The highest yields have been reached using TEBAС as phase-transfer catalyst, though the non-catalytic version proves to be also efficient (DS 2.8, 70% yield). The propargylation of AG is less effective in the systems MОН (M=Na, K)/DMSO. DS of propargyl AG reaches 1.8 (70% yield) when propargyl chloride is used as propargylating agent, while with PB under the same conditions DS value does not exceed 0.4-0.6, the yields being 44-65%. Under the action of t-BuOK/DMSO system, the AG propargyl ethers have been isomerized to the corresponding unstable allenyl ethers. AG propargyl ethers with DS 2.0 have been almost exhaustively metallated with Ag2O/NH3 system to give AG-Ag acetylides derivatives with Ag/C≡C ratio equaling 0.95.
Sigma-Aldrich
Propargyl chloride, 98%
Sigma-Aldrich
Propargyl bromide solution, purum, ~80% in toluene