- Cutting edge: NF-kappa B2 is a negative regulator of dendritic cell function.
Cutting edge: NF-kappa B2 is a negative regulator of dendritic cell function.
Journal of immunology (Baltimore, Md. : 1950) (2004-01-07)
Kendra Speirs, Linda Lieberman, Jorge Caamano, Christopher A Hunter, Phillip Scott
PMID14707043
ABSTRACT
RelB, a member of the NF-kappaB family of transcription factors, is essential for dendritic cell (DC) maturation. Recent findings indicate that RelB is exclusively regulated through its interaction with cytoplasmic NF-kappaB2/p100. The studies presented in this report show that DCs lacking NF-kappaB2 have dramatically enhanced RelB activity, associated with increased MHC class II and costimulatory molecule expression and an enhanced ability to induce CD4(+) T cell responses. These studies identify a novel role for NF-kappaB2 in the negative regulation of RelB-induced DC maturation, with critical consequences for the regulation of adaptive immune responses.
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, BioXtra, suitable for cell culture, γ-irradiated