Merck
  • Effect of vaccination and the immunomodulators "bacillus of Calmette-Guérin, avridine and Propionibacterium acnes" on rabies in mice.
Effect of vaccination and the immunomodulators "bacillus of Calmette-Guérin, avridine and Propionibacterium acnes" on rabies in mice.
Comparative immunology, microbiology and infectious diseases (1998-10-17)
J Megid, M T Peraçolli, P R Curi, C R Zanetti, W H Cabrera, R Vassao, F H Ito
ABSTRACT
Responses of vaccination and treatment to immunomodulators against rabies in mice were evaluated through macrophage inhibition factor (MIF), intra-pad inoculation (IPI) and serum neutralization (SN) tests and by the detection of gamma-interferon (IFN-gamma). Onco-BCG, Avridine and Propionibacterium acnes were administered to groups of mice. Higher survival rates were found in animals treated with P. acnes. Lower levels of IFN-gamma were observed in the groups of infected and vaccinated mice. The IPI was not effective on detecting the response of delayed-type hypersensitivity. Vaccine induced in the infected animals a more intense response to MIF reaction.
Sigma-Aldrich
Avridine, ≥97% (HPLC)