Merck
  • Defluorination of 4-fluorophenol by cytochrome P450(BM₃)-F87G: activation by long chain fatty aldehydes.
Defluorination of 4-fluorophenol by cytochrome P450(BM₃)-F87G: activation by long chain fatty aldehydes.
Biotechnology letters (2012-05-29)
Alexandria Harkey, Hye-Jin Kim, Suneel Kandagatla, Gregory M Raner
ABSTRACT
Cytochrome P450(BM3)-F87G catalyzed the oxidative defluorination of 4-fluorophenol, followed by reduction of the resulting benzoquinone to hydroquinone via the NADPH P450-reductase activity of the enzyme. The k (cat) and K (m) for this reaction were 71 ± 5 min(-1) and 9.5 ± 1.3 mM, respectively. Co-incubation of the reaction mixture with long chain aldehydes stimulated the defluorination reaction, with the 2,3-unsaturated aldehyde, 2-decenal producing a 12-fold increase in catalytic efficiency. At 150 μM aldehyde, k (cat) increased to 158 ± 4, while K (m) decreased to 1.8 ± 0.2. The effects of catalase, glutathione and ascorbate on the reaction were all consistent with a direct oxygen insertion mechanism, as opposed to a radical mechanism. The study demonstrates the potential use of P450(BM3) mutants in oxidative defluorination reactions, and characterizes the novel stimulatory action of straight chain aldehydes on this activity.
Sigma-Aldrich
4-Fluorophenol, 99%