Merck
  • Characterization and reactivity of photodimers of dihydroneopterin and dihydrobiopterin.
Characterization and reactivity of photodimers of dihydroneopterin and dihydrobiopterin.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2012-02-01)
Mariana Vignoni, Carolina Lorente, Franco M Cabrerizo, Rosa Erra-Balsells, Esther Oliveros, Andrés H Thomas
ABSTRACT
7,8-Dihydrobiopterin (H(2)Bip) and 7,8-dihydroneopterin (H(2)Nep) belong to a class of heterocyclic compounds present in a wide range of living systems. H(2)Bip accumulates in the skin of patients suffering from vitiligo, whereas H(2)Nep is secreted by human macrophages when the cellular immune system is activated. We have investigated the photochemical reactivity of both compounds upon UV-A irradiation (320-400 nm), the chemical structures of the products and their thermal stability. The study was performed in neutral aqueous solutions. The reactions were followed by UV/Visible spectrophotometry and HPLC and the products were analyzed by means of electrospray ionization mass spectrometry and (1)H-NMR. Excitation of H(2)Bip and H(2)Nep leads to the formation, in each case, of two main isomeric dimers. The latter compounds undergo a thermal process that may consist in a retro [2 + 2]-cycloaddition and hydrolysis to yield the reactant (H(2)Bip or H(2)Nep) and a product that has incorporated a molecule of H(2)O.
Sigma-Aldrich
7,8-Dihydro-L-biopterin, ≥94% (HPLC)
Sigma-Aldrich
7,8-Dihydroneopterin, ≥97.0% (HPLC)