Merck
  • Comb-shaped glycopolymer/peptide bioconjugates by combination of RAFT polymerization and thiol-ene "click" chemistry.
Comb-shaped glycopolymer/peptide bioconjugates by combination of RAFT polymerization and thiol-ene "click" chemistry.
Macromolecular bioscience (2012-10-09)
Xiaobei Wang, Li Liu, Yan Luo, Haiting Shi, Jingyi Li, Hanying Zhao
ABSTRACT
Comb-shaped glycopolymer/peptide bioconjugates are constructed by grafting reduced glutathione (GSH) onto acrylate-functional block glycocopolymers via thiol-ene click chemistry. In aqueous solution, the glycopolymer/GSH bioconjugate self-assembles to sugar-installed spherical micelles. The size of micelles decreases with increasing pH, demonstrating pH-responsive character. The isoelectric point (IEP) of the PMAGlc/GSH bioconjugate is estimated to be 3.43. The micelles show a specific interaction with the protein Concanavalin A. At endosomal pH, the PMAGlc/GSH bioconjugate can gradually degrade. These pH-responsive glycopolymer/peptide micelles with biological recognition and degradation can be used as multifunctional nanocarriers for targeted drug delivery.
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Zinc acrylate, 98%