Merck
  • Inhibitors of acyl-CoA:cholesterol O-acyltransferase. 17. Structure-activity relationships of several series of compounds derived from N-chlorosulfonyl isocyanate.
Inhibitors of acyl-CoA:cholesterol O-acyltransferase. 17. Structure-activity relationships of several series of compounds derived from N-chlorosulfonyl isocyanate.
Journal of medicinal chemistry (1996-03-15)
J A Picard, P M O'Brien, D R Sliskovic, M K Anderson, R F Bousley, K L Hamelehle, B R Krause, R L Stanfield
ABSTRACT
Several series of acyl-CoA:cholesterol O-acyltransferase inhibitors were prepared by the stepwise addition of nitrogen, oxygen, and sulfur nucleophiles to N-chlorosulfonyl isocyanate. The (aminosulfonyl)ureas 3-44 were the most potent inhibitors in vitro, with several compounds having IC50 values < 1 microM. Although the other series of compounds were not as potent in vitro, many compounds did display good in vivo activity in cholesterol-fed rats. Several of the oxysulfonyl carbamates (including CI-999, 115) showed excellent lipid-lowering activity in the chronic in vivo screen, demonstrating significant cholesterol lowering in a pre-established hypercholesterolemic state.
Sigma-Aldrich
Chlorosulfonyl isocyanate, 98%
Sigma-Aldrich
Chlorosulfonyl isocyanate, Arxada quality, 99.0-100.3 % (w/w) (T)