Merck
  • Complete sets of monosubstituted γ-cyclodextrins as precursors for further synthesis.
Complete sets of monosubstituted γ-cyclodextrins as precursors for further synthesis.
The Journal of organic chemistry (2012-12-05)
Markéta Bláhová, Eva Bednářová, Michal Řezanka, Jindřich Jindřich
ABSTRACT
Regioselective alkylation of γ-cyclodextrin with allyl or propargyl bromide, using optimized reaction conditions, followed by peracetylation of the remaining hydroxyl groups and separation of isomers resulted in the set of peracetylated 2(I)-O-, 3(I)-O- and 6(I)-O-alkylated cyclodextrins in up to 19% yields. Ozonolysis or oxidative cleavage of peracetylated allyl derivatives resulted in a complete set of peracetylated 2(I)-O-, 3(I)-O-, and 6(I)-O-formylmethyl or -carboxymethyl derivatives. All of these derivatives are useful precursors for further preparation of regioselectively monosubstituted derivatives of γ-cyclodextrin.
Sigma-Aldrich
N-Methyl-N-propargylbenzylamine, 97%
Sigma-Aldrich
Pargyline hydrochloride