Merck
Bi-doped BaF2 crystal for broadband near-infrared light source.
Optics express (2009-04-01)
Jian Ruan, Liangbi Su, Jianrong Qiu, Danping Chen, Jun Xu
ABSTRACT
Bi-doped BaF(2) crystal was grown by the temperature gradient technique and its spectral properties were investigated. The absorption, emission and excitation spectra were measured at room temperature. Two broadband emissions centered at 1070 and 1500 nm were observed in Bi-doped BaF(2) crystal. This extraordinary luminescence should be ascribed to Bi-related centers at distinct sites. We suggest Bi(2+) or Bi(+) centers adjacent to F vacancy defects are the origins of the observed NIR emissions.
Sigma-Aldrich
Barium fluoride, precipitated, 99.95% trace metals basis
Sigma-Aldrich
Barium fluoride, powder, 98%
Sigma-Aldrich
Barium fluoride, 99.99% trace metals basis