Merck
  • A chiral HPLC method for the determination of low amounts of D-carnitine in L-carnitine after derivatization with (+)-FLEC.
A chiral HPLC method for the determination of low amounts of D-carnitine in L-carnitine after derivatization with (+)-FLEC.
Journal of pharmaceutical and biomedical analysis (2002-08-23)
Sascha Freimüller, Hans Altorfer
ABSTRACT
An indirect enantioseparation method for robust and precise determination of D-Carnitine (D-C) in L-Carnitine (L-C) in the range of 0.1-1.0% is presented. The method is based on derivatization of Carnitine with (+)-[1-(9-fluorenyl)-ethyl]-chloroformate ((+)-FLEC). The two diastereomers are subsequently separated of on an octadecyl column using detection of the eluent by fluorescence (260 excitation, 310 nm emission monitoring). This procedure can be calibrated conveniently by diluting the derivatization solution of the sample. Hence, D-C was determined indirectly through quantification of L-C thereby strongly increasing the robustness and reducing the costs. During the development of the method a study was undertaken to prove that the method is suitable to determine enantiomeric purity of L-C indeed. Moreover, the method was validated according to the ICH guidance, which required the additional performance of a collaborative study. The proposed assay can be carried out using an autoinjector because the derivatives are very stable. Hence, we believe that this method will become popular for reliable determination of enantiomeric purity of L-C.
Product Description
Supelco
(+)-1-(9-Fluorenyl)ethyl chloroformate solution, ≥18 mM in acetone, for chiral derivatization, LiChropur
Sigma-Aldrich
(+)-1-(9-Fluorenyl)ethyl chloroformate solution, 18 mM in acetone, for chiral derivatization