Merck
  • CD16+ monocytes in breast cancer patients: expanded by monocyte chemoattractant protein-1 and may be useful for early diagnosis.
CD16+ monocytes in breast cancer patients: expanded by monocyte chemoattractant protein-1 and may be useful for early diagnosis.
Clinical and experimental immunology (2011-03-03)
A-L Feng, J-K Zhu, J-T Sun, M-X Yang, M R Neckenig, X-W Wang, Q-Q Shao, B-F Song, Q-F Yang, B-H Kong, X Qu
ABSTRACT
Human peripheral blood monocytes are a heterogeneous population, including CD14(+) CD16(-) 'classical' monocytes and CD14(+) CD16(+) 'proinflammatory' monocytes. CD16(+) monocytes are expanded in various inflammatory conditions. However, little is known about the CD14(+) CD16(+) monocytes in patients with breast cancer. We detected CD14(+) CD16(+) monocytes in 96 patients with breast cancer and 54 control subjects using flow cytometry. Receiver-operating characteristic (ROC) curve analysis was used to determine the feasibility of CD14(+) CD16(+) monocytes as an indicator for diagnosis of breast cancer. We found that the frequency of CD14(+) CD16(+) monocytes showed a significantly greater increase in breast cancer patients than in controls (16·96% versus 10·84%, P < 0·0001). The area under the ROC curve for CD14(+) CD16(+) monocytes was 0·805 [95% confidence interval (95% CI): 0·714-0·877, P = 0·0001]. Furthermore, the levels of CD16(+) monocytes were significantly negatively associated with the tumour size and pathological staging. In vitro, we showed that CD14(+) CD16(+) monocytes were expanded significantly when the purified CD14(+) monocytes were exposed to Michigan Cancer Foundation (MCF)-7 cells-conditioned medium (MCF-CM) or, separately, to monocyte chemotactic protein 1 (MCP-1). Neutralizing antibodies against MCP-1 inhibited the expansion of CD14(+) CD16(+) monocytes by MCF-CM. Collectively, our findings indicated that MCP-1 can expand CD14(+) CD16(+) monocytes in patients with breast cancer. Furthermore, the CD14(+) CD16(+) monocyte may be a useful indicator in early diagnosis of breast cancer.
Sigma-Aldrich
CD14 human, recombinant, expressed in E. coli, 0.5 mg protein/mL
Pricing and availability is not currently available.