Merck
  • The GSK3β-β-catenin-TCF1 pathway improves naive T cell activation in old adults by upregulating miR-181a.
The GSK3β-β-catenin-TCF1 pathway improves naive T cell activation in old adults by upregulating miR-181a.
NPJ aging and mechanisms of disease (2021-02-10)
Zhongde Ye, Timothy M Gould, Huimin Zhang, Jun Jin, Cornelia M Weyand, Jörg J Goronzy
ABSTRACT
MicroRNAs play an important role in the regulation of T cell development, activation, and differentiation. One of the most abundant microRNAs in lymphocytes is miR-181a, which controls T cell receptor (TCR) activation thresholds in thymic selection as well as in peripheral T cell responses. We previously found that miR-181a levels decline in T cells in the elderly. In this study, we identified TCF1 as a transcriptional regulator of pri-miR-181a. A decline in TCF1 levels in old individuals accounted for the reduced miR-181a expression impairing TCR signaling. Inhibition of GSK3ß restored expression of miR-181a by inducing TCF1 in T cells from old adults. GSK3ß inhibition enhanced TCR signaling to increase downstream expression of activation markers and production of IL-2. The effect involved the upregulation of miR-181a and the inhibition of DUSP6 expression. Thus, inhibition of GSK3ß can restore responses of old T cells by inducing miR-181a expression through TCF1.
Sigma-Aldrich
SB 216763, >98% (HPLC)
Sigma-Aldrich
GSK-3 Inhibitor IX, BIO, CAS 667463-62-9, is a cell-permeable, highly potent, selective, reversible, and ATP-competitive inhibitor of GSK-3α/β (IC₅₀ = 5 nM). Maintains self-renewal in human & mouse embryonic stem cells.