Merck
  • The N-terminal Leu-Pro-Gln sequence of Rab34 is required for ciliogenesis in hTERT-RPE1 cells.
The N-terminal Leu-Pro-Gln sequence of Rab34 is required for ciliogenesis in hTERT-RPE1 cells.
Small GTPases (2021-04-17)
Mai E Oguchi, Yuta Homma, Mitsunori Fukuda
ABSTRACT
We have previously shown that Rab34 is an important regulator of ciliogenesis and that its unique long N-terminal region (amino acids 1-49) is essential for ciliogenesis in certain cultured mammalian cells. In the present study, we performed an in-depth deletion analysis of the N-terminal region of Rab34 together with Ala-based site-directed mutagenesis to identify the essential amino acids that are required for serum-starvation-induced ciliogenesis in hTERT-RPE1 cells. The results showed that a Rab34 mutant lacking an N-terminal 18 amino acids and a Rab34 mutant carrying an LPQ-to-AAA mutation (amino acids 16-18) failed to rescue a Rab34-KO phenotype (i.e., defect in ciliogenesis). Our findings suggest that the LPQ sequence of Rab34 is crucial for ciliogenesis in hTERT-RPE1 cells.Abbreviations: AA, amino acid(s); ac-Tub, acetylated tubulin; bsr, blasticidin S-resistant gene; HRP, horseradish peroxidase; hTERT-RPE1, human telomerase reverse transcriptase retinal pigment epithelium 1; KO, knockout; NS, not significant; PBS, phosphate-buffered saline; puro, puromycin-resistant gene.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-GALNT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution