Merck
  • Effect of beta-alanine supplementation on muscle carnosine concentrations and exercise performance.
Effect of beta-alanine supplementation on muscle carnosine concentrations and exercise performance.
Amino acids (2010-01-22)
Craig Sale, Bryan Saunders, Roger C Harris
ABSTRACT
High-intensity exercise results in reduced substrate levels and accumulation of metabolites in the skeletal muscle. The accumulation of these metabolites (e.g. ADP, Pi and H(+)) can have deleterious effects on skeletal muscle function and force generation, thus contributing to fatigue. Clearly this is a challenge to sport and exercise performance and, as such, any intervention capable of reducing the negative impact of these metabolites would be of use. Carnosine (beta-alanyl-L-histidine) is a cytoplasmic dipeptide found in high concentrations in the skeletal muscle of both vertebrates and non-vertebrates and is formed by bonding histidine and beta-alanine in a reaction catalysed by carnosine synthase. Due to the pKa of its imidazole ring (6.83) and its location within skeletal muscle, carnosine has a key role to play in intracellular pH buffering over the physiological pH range, although other physiological roles for carnosine have also been suggested. The concentration of histidine in muscle and plasma is high relative to its K (m) with muscle carnosine synthase, whereas beta-alanine exists in low concentration in muscle and has a higher K (m) with muscle carnosine synthase, which indicates that it is the availability of beta-alanine that is limiting to the synthesis of carnosine in skeletal muscle. Thus, the elevation of muscle carnosine concentrations through the dietary intake of carnosine, or chemically related dipeptides that release beta-alanine on absorption, or supplementation with beta-alanine directly could provide a method of increasing intracellular buffering capacity during exercise, which could provide a means of increasing high-intensity exercise capacity and performance. This paper reviews the available evidence relating to the effects of beta-alanine supplementation on muscle carnosine synthesis and the subsequent effects on exercise performance. In addition, the effects of training, with or without beta-alanine supplementation, on muscle carnosine concentrations are also reviewed.
Sigma-Aldrich
β-Alanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
β-Alanine, 99%
Sigma-Aldrich
β-Alanine, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
β-Alanine, BioXtra, ≥99.0% (NT)
Supelco
β-Alanine, analytical standard
Supelco
Beta Alanine, Pharmaceutical Secondary Standard; Certified Reference Material