Merck
  • Perlecan is recruited by dystroglycan to nodes of Ranvier and binds the clustering molecule gliomedin.
Perlecan is recruited by dystroglycan to nodes of Ranvier and binds the clustering molecule gliomedin.
The Journal of cell biology (2015-02-04)
Cristina Colombelli, Marilena Palmisano, Yael Eshed-Eisenbach, Desirée Zambroni, Ernesto Pavoni, Cinzia Ferri, Stefania Saccucci, Sophie Nicole, Raija Soininen, Karen K McKee, Peter D Yurchenco, Elior Peles, Lawrence Wrabetz, M Laura Feltri
ABSTRACT
Fast neural conduction requires accumulation of Na(+) channels at nodes of Ranvier. Dedicated adhesion molecules on myelinating cells and axons govern node organization. Among those, specific laminins and dystroglycan complexes contribute to Na(+) channel clustering at peripheral nodes by unknown mechanisms. We show that in addition to facing the basal lamina, dystroglycan is found near the nodal matrix around axons, binds matrix components, and participates in initial events of nodogenesis. We identify the dystroglycan-ligand perlecan as a novel nodal component and show that dystroglycan is required for the selective accumulation of perlecan at nodes. Perlecan binds the clustering molecule gliomedin and enhances clustering of node of Ranvier components. These data show that proteoglycans have specific roles in peripheral nodes and indicate that peripheral and central axons use similar strategies but different molecules to form nodes of Ranvier. Further, our data indicate that dystroglycan binds free matrix that is not organized in a basal lamina.
Sigma-Aldrich
Monoclonal Anti-Sodium Channel, Pan antibody produced in mouse, ~1 mg/mL, clone K58/35, purified immunoglobulin
Sigma-Aldrich
Monoclonal Anti-Dystrophin antibody produced in mouse, clone MANDRA1, ascites fluid
Sigma-Aldrich
Complement, Rabbit Serum, Complement, Rabbit Serum, is tested for suitability for HLA-ABC serology. Useful as an active source of complement for use with hybridoma antibodies, alloantisera, and related applications.
Sigma-Aldrich
Anti-Heparan Sulfate Proteoglycan (Perlecan) Antibody, clone A7L6, clone A7L6, from rat