Merck
  • MiR-148a-3p Regulates the Invasion and Odontoblastic Differentiation of Human Dental Pulp Stem Cells via the Wnt1/β-Catenin Pathway.
MiR-148a-3p Regulates the Invasion and Odontoblastic Differentiation of Human Dental Pulp Stem Cells via the Wnt1/β-Catenin Pathway.
International journal of stem cells (2021-08-31)
Qiong Li, Lei Huang
ABSTRACT
MiR-148a-3p has been reported to regulate the differentiation of marrow stromal cell osteoblast. In this study, whether miR-148a-3p regulated the odontoblastic differentiation of human dental pulp stem cells (hDPSCs) or not was explored. The hDPSCs were isolated and identified via flow cytometry. Targets of miR-148a-3p were identified via bioinformatics and dual-luciferase reporter assay. After the cell was cultured in the odontogenic differentiation medium or infected, cell viability, invasion, and odontoblastic differentiation were detected via MTT, transwell, and Alizarin Red S staining, respectively. The miR-148a-3p, Wnt1, β-catenin, DSPP, DMP-1, RUNX2, OCN, and Smad4 expressions were determined by RT-qPCR and Western blot. The hDPSCs odontoblastic differentiation downregulated the miR-148a-3p expression and upregulated Wnt1 expression. Wnt1 was determined as the target for miR-148a-3p. MiR-148a-3p mimic and siWnt1 suppressed the cell viability, invasion, and odontoblastic differentiation of hDPSCs and inhibited the Wnt1, β-catenin, DSPP, DMP-1, RUNX2, OCN, and Smad4 expressions. In contrast, miR-148a-3p inhibitor and overexpressed Wnt1 promoted the cell viability, invasion, and odontoblastic differentiation of hDPSCs, and upregulated the Wnt1, β-catenin, DSPP, DMP-1, RUNX2, OCN, and Smad4 expressions. Also, miR-148a-3p mimic and inhibitor reversed the effects of Wnt1 overexpression and siWnt1. MiR-148a-3p modulated the invasion and odontoblastic differentiation of hDPSCs through the Wnt1/β-catenin pathway.
Sigma-Aldrich
ChemiScreen Recombinant Human Glucagon Receptor Membrane Preparation, Human glucagon / GCG GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Sigma-Aldrich
ANTI-DSPP (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution