Merck
  • Transcuprein is a macroglobulin regulated by copper and iron availability.
Transcuprein is a macroglobulin regulated by copper and iron availability.
The Journal of nutritional biochemistry (2007-03-17)
Nanmei Liu, Louis Shi-li Lo, S Hassan Askary, LaTrice Jones, Theodros Z Kidane, Trisha Trang, Minh Nguyen, Jeremy Goforth, Yu-Hsiang Chu, Esther Vivas, Monta Tsai, Terence Westbrook, Maria C Linder
ABSTRACT
Transcuprein is a high-affinity copper carrier in the plasma that is involved in the initial distribution of copper entering the blood from the digestive tract. To identify and obtain cDNA for this protein, it was purified from rat plasma by size exclusion and copper-chelate affinity chromatography, and amino acid sequences were obtained. These revealed a 190-kDa glycosylated protein identified as the macroglobulin alpha(1)-inhibitor III, the main macroglobulin of rodent blood plasma. Albumin (65 kDa) copurified in variable amounts and was concluded to be a contaminant (although it can transiently bind the macroglobulin). The main macroglobulin in human blood plasma (alpha(2)-macroglobulin), which is homologous to alpha(1)-inhibitor III, also bound copper tightly. Expression of alpha(1)I3 (transcuprein) mRNA by the liver was examined in rats with and without copper deficiency, using quantitative polymerase chain reaction methodology and Northern blot analysis. Protein expression was examined by Western blotting. Deficient rats with 40% less ceruloplasmin oxidase activity and liver copper concentrations expressed about twice as much alpha(1)I3 mRNA, but circulating levels of transcuprein did not differ. Iron deficiency, which increased liver copper concentrations by threefold, reduced transcuprein mRNA expression and circulating levels of transcuprein relative to what occurred in rats with normal or excess iron. We conclude that transcupreins are specific macroglobulins that not only carry zinc but also carry transport copper in the blood, and that their expression can be modulated by copper and iron availability.
Product Description
Millipore
Protein A–Agarose macrobeads, aqueous suspension
Millipore
Protein A–Agarose, saline suspension