
Voriconazole Related Compound C

Synonym(s):
2,4-Difluoro-α-(1H-1,2,4-triazolyl)acetophenone, 1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone
Empirical Formula (Hill Notation):
C10H7F2N3O
CAS Number:
Molecular Weight:
223.18
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing