
Trimebutine maleate

Synonym(s):
Trimebutine maleate salt, 3,4,5-Trimethoxybenzoic acid 2-(dimethylamino)-2-phenylbutyl ester
Empirical Formula (Hill Notation):
C22H29NO5 · C4H4O4
CAS Number:
Molecular Weight:
503.54
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2