
Sulfamethoxazole Related Compound C

Synonym(s):
3-Amino-5-methylisoxazole, 5-Methyl-3-isoxazolamine, NSC 159134
Empirical Formula (Hill Notation):
C4H6N2O
CAS Number:
Molecular Weight:
98.10
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2