
Sulfamethoxazole Related Compound B

Synonym(s):
4-Amino-N-{4-[N-(5-methylisoxazol-3-yl)sulfamoyl]phenyl}benzenesulfonamide
Empirical Formula (Hill Notation):
C16H16N4O5S2
CAS Number:
Molecular Weight:
408.45
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2