
Solasodine

Synonym(s):
(-)-Solasodine, (3β,22α,25R)-Spirosol-5-en-3-ol, NSC 178260, NSC 179187, Purapuridine, Solancarpidine, Solasod-5-en-3β-ol, Solasod-5-en-3β-ol (7CI,8CI), Solasodin
Empirical Formula (Hill Notation):
C27H43NO2
CAS Number:
Molecular Weight:
413.64
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing