Skip to Content
Merck
Seleno-L-cystine 95%

Seleno-L-cystine

Seleno-L-cystine 95%
Synonym(s):
(R,R)-3,3′-Diseleno-bis(2-aminopropionic acid), L-Selenocystine
Linear Formula:
CO2HCH(NH2)CH2(Se)2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
334.09

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing