
RoflumilastRelated Compound D

Synonym(s):
3-(Cyclopropylmethoxy)-4-(difluoromethoxy)benzoic acid
Empirical Formula (Hill Notation):
C12H12F2O4
CAS Number:
Molecular Weight:
258.22
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing