
Pyrex® disposable culture tubes, screw-cap style, flat bottom

Synonym(s):
Corning® 99448
Products
Product Category
Color
Manufacturer
Binding Type
Capacity (mL)
OD (mm)
Cap
Package Size
Sterility
Showing 1-3 of 3
Compare
Product Number
Description
Pricing