
Pravastatin Related Compound B

Synonym(s):
(βR,δR,1S,2S,6R,8S,8aR)-1,2,6,7,8,8a-Hexahydro-β,δ,6-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-1-naphthaleneheptanoic acid monosodium salt, 6-epi-Pravastatin sodium salt
Empirical Formula (Hill Notation):
C23H35NaO7
CAS Number:
Molecular Weight:
446.51
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing