Skip to Content
Merck
Poloxamer 124 European Pharmacopoeia (EP) Reference Standard

Poloxamer 124

Poloxamer 124 European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)
CAS Number:

Products

Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing