
Phloroglucinol (anhydrous)

Synonym(s):
Phloroglucinol, 1,3,5-Trihydroxybenzene
Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing