
Phenylcyclohexylglycolic acid

Synonym(s):
α-Cyclohexylmandelic acid, 2-Cyclohexyl-2-hydroxy-2-phenylacetic acid
Empirical Formula (Hill Notation):
C14H18O3
CAS Number:
Molecular Weight:
234.29
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing