
Olopatadine Related Compound C

Synonym(s):
(11-Oxo-6,11-dihydrodibenzo[b,e]oxepin-2-yl)acetic acid, Isoxepac, NSC 300907
Empirical Formula (Hill Notation):
C16H12O4
Molecular Weight:
268.26
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2