Monoclonal Anti-Cytokeratin Peptide 18−FITC antibody produced in mouse
Synonym(s):
Monoclonal Anti-Cytokeratin Peptide 18
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing