
L-Alanyl-L-glutamine

Synonym(s):
Ala-Gln, Alanyl-glutamine, Glutamine-S
Empirical Formula (Hill Notation):
C8H15N3O4
CAS Number:
Molecular Weight:
217.22
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2