
Isoflurane

Synonym(s):
Isoflurane, 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether
Empirical Formula (Hill Notation):
C3H2ClF5O
CAS Number:
Molecular Weight:
184.49
Products
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Functional Group
Application
Package Size
API Family
Reference Material Type
Special Grade