
Ibuprofen impurity F

Synonym(s):
3-(4-Isobutylphenyl)propionic acid
Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2