
Hexane-1-sulfonic acid sodium salt

Synonym(s):
Hexane-1-sulfonic acid sodium salt
Empirical Formula (Hill Notation):
C6H13NaO3S
CAS Number:
Molecular Weight:
188.22
Products
Brand
Formula Weight
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing